2,3-Dimethyl-2,3-dinitrobutane ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Pericolo
|
D1822-10G
D1822-25G
TCIAD1822-10GEA
53
EUR
TCIAD1822-10G
TCIAD1822-25G
2,3-Dimethyl-2,3-dinitrobutane ≥98.0% (by GC)
2,3-Dimethyl-2,3-dinitrobutane
Formula:
C₆H₁₂N₂O₄ Peso molecolare: 176,17 g/mol Punto di fusione: 215°C Densità: 1,34 g/cm³ (20 °C) |
MDL Number:
MFCD00007392 CAS Number: 3964-18-9 EINECS: 223-569-9 UN: 2926 ADR: 4.1,III |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...