Isopentylamine ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
1-Amino-3-methylbutane, Isopentyl amine, Isoamylamine, 3-Methylbutylamine, Leucamine
I0082-25ML
I0082-500ML
I0082-100ML
TCIAI0082-25MLEA
18
EUR
TCIAI0082-25ML
TCIAI0082-500ML
TCIAI0082-100ML
Isopentylamine ≥98.0% (by GC, titration analysis)
Isopentylamine
Formula:
C₅H₁₃N Peso molecolare: 87,16 g/mol Punto di ebollizione: 97°C Punto di fusione: –68 °C Densità: 0,75 g/cm³ (20 °C) Flash Pt: -1°C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00008203 CAS Number: 107-85-7 EINECS: 203-526-0 UN: 1106 ADR: 3,II Merck Index: 14,05112 |
Risultati del test di specificazione
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...