cis-3-Hexene ≥97.0% (by GC)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
(Z)-3-Hexene
H0398-1ML
H0398-5ML
TCIAH0398-1MLEA
16
EUR
TCIAH0398-1ML
TCIAH0398-5ML
cis-3-Hexene ≥97.0% (by GC)
cis-3-Hexene
Formula:
C₆H₁₂ Peso molecolare: 84,16 g/mol Punto di ebollizione: 67°C Flash Pt: –25°C |
MDL Number:
MFCD00085300 CAS Number: 7642-09-3 UN: 3295 ADR: 3,II |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...