2-(Dicyclohexylphosphino)biphenyl ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
(2-Biphenyl)dicyclohexylphosphine
D3388-1G
D3388-5G
TCIAD3388-1GEA
33
EUR
TCIAD3388-1G
TCIAD3388-5G
2-(Dicyclohexylphosphino)biphenyl ≥98.0% (by GC)
2-(Dicyclohexylphosphino)biphenyl
Formula:
C₂₄H₃₁P Peso molecolare: 350,48 g/mol |
MDL Number:
MFCD01862441 CAS Number: 247940-06-3 EINECS: 480-030-2 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 101.0 to 105.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...