Potassium(4-methoxyphenyl)trifluoroborate ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
(4-Methoxyphenyl)trifluoroboric acid potassium salt, Potassium (4-methoxyphenyl)trifluoroborate, Potassium trifluoro(4-methoxyphenyl)borate
P1820-1G
P1820-5G
TCIAP1820-1GEA
38
EUR
TCIAP1820-1G
TCIAP1820-5G
Potassium(4-methoxyphenyl)trifluoroborate ≥98.0% (by HPLC, titration analysis)
Potassium(4-methoxyphenyl)trifluoroborate
Formula:
C₇H₇BF₃KO Peso molecolare: 214,04 g/mol |
MDL Number:
MFCD04112717 CAS Number: 192863-36-8 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Ion exchange titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...