Potassium(3-methoxyphenyl)trifluoroborate ≥98.0% (by HPLC)
Fornitore: TCI
|
Attenzione
|
P1580-1G
P1580-5G
TCIAP1580-1GEA
36
EUR
TCIAP1580-1G
TCIAP1580-5G
Potassium(3-methoxyphenyl)trifluoroborate ≥98.0% (by HPLC)
Potassium(3-methoxyphenyl)trifluoroborate
Formula:
C₇H₇BF₃KO |
MDL Number:
MFCD04039954 CAS Number: 438553-44-7 |
|
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...