Stampa…

Perfluoroheptanoic acid ≥98.0% (by titrimetric analysis)

Fornitore: TCI
Pericolo

T1545-25G T1545-5G
TCIAT1545-25GEA 193 EUR
TCIAT1545-25G TCIAT1545-5G
Perfluoroheptanoic acid ≥98.0% (by titrimetric analysis)
Perfluoroheptanoic acid
Formula: C₇HF₁₃O₂
Punto di ebollizione: 177°C
MDL Number: MFCD00039604
CAS Number: 375-85-9
EINECS: 206-798-9
UN: 3261
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c3e72b1766f372d146f8aa2235c8e95e59e597cf&vid=c3e72b1766f372d146f8aa2235c8e95e59e597cf&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR