4'-Bromo-3'-nitroacetophenone ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
1-(4-Bromo-3-nitrophenyl)ethan-1-one, 1-(4-Bromo-3-nitrophenyl)ethanone
B3383-1G
B3383-5G
TCIAB3383-1GEA
16
EUR
TCIAB3383-1G
TCIAB3383-5G
4'-Bromo-3'-nitroacetophenone ≥98.0% (by GC)
4'-Bromo-3'-nitroacetophenone
Formula:
C₈H₆BrNO₃ Peso molecolare: 244,04 g/mol Punto di fusione: 120 °C |
MDL Number:
MFCD00016985 CAS Number: 18640-58-9 EINECS: 242-469-6 |
Risultati del test di specificazione
Appearance | Light yellow to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 117.0 to 121.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...