Stampa…

Sodium methyl sulphate ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Methanesulphuric acid sodium salt, Methylsulphuric acid sodium salt

M0433-25G M0433-250G
TCIAM0433-25GEA 92 EUR
TCIAM0433-25G TCIAM0433-250G
Sodium methyl sulphate ≥98.0% (by titrimetric analysis)
Sodium methyl sulphate
Formula: CH₃NaO₄S
Peso molecolare: 134,09 g/mol
Punto di fusione: 193 °C
MDL Number: MFCD00013457
CAS Number: 512-42-5
EINECS: 208-142-7
Merck Index: 14,06122
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5f84e0eb0b01cc8731a2041b23e66010a2b49c3e&vid=149338c191a9ffcbffcea1770b7d44f0bc87a5fc&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Ion exchange titration) min. 98.0 %
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR