Hexyl isocyanate ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
Isocyanic acid hexyl ester
I0492-25ML
I0492-5ML
TCIAI0492-25MLEA
194
EUR
TCIAI0492-25ML
TCIAI0492-5ML
Hexyl isocyanate ≥98.0% (by GC)
Hexyl isocyanate
Formula:
C₇H₁₃NO Peso molecolare: 127,19 g/mol Punto di ebollizione: 164°C Densità: 1,2 g/cm³ (20 °C) Flash Pt: 60°C Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00037047 CAS Number: 2525-62-4 EINECS: 219-763-8 UN: 3080 ADR: 6.1,II |
Risultati del test di specificazione
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...