Cerotic acid ≥95.0% (by GC, titration analysis)
Fornitore: TCI
C0829-1G
C0829-10G
TCIAC0829-1GEA
142
EUR
TCIAC0829-1G
TCIAC0829-10G
Cerotic acid ≥95.0% (by GC, titration analysis)
Cerotic acid
Formula:
C₂₆H₅₂O₂ Punto di ebollizione: 419 °C (1013 hPa) Punto di fusione: 76…77 °C Densità: 0,8198 g/cm³ (100 °C) Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00002811 CAS Number: 506-46-7 EINECS: 208-040-2 |
|
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 95.0 % |
Purity (Neutralization titration) | min. 95.0 % |
Melting point | 85.0 to 90.0 deg-C |
Solubility in hot Toluene | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...