Stampa…

Heneicosanoic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI
Attenzione

Synonyms: Henicosanoic acid

H0010-1G H0010-10G
TCIAH0010-1GEA 77 EUR
TCIAH0010-1G TCIAH0010-10G
Heneicosanoic acid ≥98.0% (by GC, titration analysis)
Heneicosanoic acid
Formula: C₂₁H₄₂O₂
Peso molecolare: 326.56 g/mol
Punto di ebollizione: 171°C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00002805
CAS Number: 2363-71-5
EINECS: 219-113-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3a7ee081f27ef92bd110623b69654717c8572c76&vid=adda898afa791c84b1ae6e8a1e4b6295b1ffc5f0&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 73.0 to 77.0 deg-C
Solubility in hot Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR