Stampa…

Acido laurico ≥98.0% (by GC, titration analysis)

Fornitore: TCI
Pericolo

Synonyms: Dodecanoic acid, Dodecylic acid

L0011-25G L0011-500G
TCIAL0011-25GEA 18 EUR
TCIAL0011-25G TCIAL0011-500G
Acido laurico ≥98.0% (by GC, titration analysis)
Acido laurico
Formula: C₁₂H₂₄O₂
Peso molecolare: 200.32 g/mol
Punto di ebollizione: 298°C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00002736
CAS Number: 143-07-7
EINECS: 205-582-1
Merck Index: 14,05384
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d5ef9d55108db0639b8795cd21f9664b09caed24&vid=6925feffc25737ccc153e1117bdc98408d3ea96e&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 43.0 to 46.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR