11-Heneicosanone ≥95.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
Didecyl ketone
H0532-25G
H0532-5G
TCIAH0532-25GEA
474
EUR
TCIAH0532-25G
TCIAH0532-5G
11-Heneicosanone ≥95.0% (by GC)
11-Heneicosanone
Formula:
C₂₁H₄₂O Peso molecolare: 310,56 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00048410 CAS Number: 19781-72-7 EINECS: 243-307-7 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 95.0 % |
Melting point | 63.0 to 67.0 deg-C |
Solubility in Toluene | very faint turbidity |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...