Stampa…

4-Nitrobenzenesulphonamide ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

N0697-25G N0697-5G
TCIAN0697-25GEA 114 EUR
TCIAN0697-25G TCIAN0697-5G
4-Nitrobenzenesulphonamide ≥98.0% (by HPLC, titration analysis)
4-Nitrobenzenesulphonamide
Formula: C₆H₆N₂O₄S
Peso molecolare: 202,19 g/mol
Punto di fusione: 187 °C
MDL Number: MFCD00007937
CAS Number: 6325-93-5
EINECS: 228-691-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=95c6ce21ff2da9b286d932a03a7c2fa830f49914&vid=95c6ce21ff2da9b286d932a03a7c2fa830f49914&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 180.0 to 184.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR