Stampa…

3-Nitrobenzenesulphonamide ≥98.0% (by HPLC, total nitrogen)

Fornitore: TCI EUROPE
Attenzione

N0696-25G N0696-5G
TCIAN0696-25GEA 50 EUR
TCIAN0696-25G TCIAN0696-5G
3-Nitrobenzenesulphonamide ≥98.0% (by HPLC, total nitrogen)
3-Nitrobenzenesulphonamide
Formula: C₆H₆N₂O₄S
Peso molecolare: 202,19 g/mol
Punto di fusione: 167 °C
MDL Number: MFCD00007935
CAS Number: 121-52-8
EINECS: 204-477-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=99d2905bd41c72831134796221cab4a23a89180f&vid=99d2905bd41c72831134796221cab4a23a89180f&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR