Stampa…

2-Nitrobenzenesulphonamide ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI
Attenzione

N0705-25G N0705-5G
TCIAN0705-25GEA 119 EUR
TCIAN0705-25G TCIAN0705-5G
2-Nitrobenzenesulphonamide ≥98.0% (by HPLC, titration analysis)
2-Nitrobenzenesulphonamide
Formula: C₆H₆N₂O₄S
MDL Number: MFCD00009807
CAS Number: 5455-59-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f4dd3c6d82cec4eaec7f98cf0cc2b0af9cd3e2d5&vid=f4dd3c6d82cec4eaec7f98cf0cc2b0af9cd3e2d5&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 192.0 to 196.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR