4-Methyl-3-nitrobenzenesulphonyl chloride ≥98.0% (by GC, titration analysis)
Fornitore: TCI
|
Pericolo
|
Formula:
C₇H₆ClNO₄S Punto di ebollizione: 154°C |
MDL Number:
MFCD00129811 CAS Number: 616-83-1 UN: 3261 ADR: 8,III |
|
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Melting point | 32.0 to 36.0 deg-C |
Solubility in Toluene | very faint turbidity |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...