2-Methyl-5-nitrobenzenesulphonyl chloride ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
4-Nitrotoluene-2-sulphonyl chloride, 2-Methyl-5-nitrobenzene-1-sulphonyl chloride
M2178-25G
M2178-5G
TCIAM2178-25GEA
212
EUR
TCIAM2178-25G
TCIAM2178-5G
2-Methyl-5-nitrobenzenesulphonyl chloride ≥98.0% (by GC, titration analysis)
2-Methyl-5-nitrobenzenesulphonyl chloride
Formula:
C₇H₆ClNO₄S Peso molecolare: 235,65 g/mol Punto di ebollizione: 198°C Punto di fusione: 46 °C |
MDL Number:
MFCD00051695 CAS Number: 121-02-8 EINECS: 204-444-8 UN: 3261 ADR: 8,III |
Risultati del test di specificazione
Appearance | White to Almost white powder to lump |
Purity (GC) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Melting point | 44.0 to 48.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...