2-Mercapto-5-nitrobenzimidazole ≥98.0% (by HPLC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
5-Nitrobenzimidazole-2-thiol, 2-Mercapto-5-nitro-1H-benzimidazole
M1325-25G
M1325-5G
TCIAM1325-25GEA
488
EUR
TCIAM1325-25G
TCIAM1325-5G
2-Mercapto-5-nitrobenzimidazole ≥98.0% (by HPLC)
2-Mercapto-5-nitrobenzimidazole
Formula:
C₇H₅N₃O₂S Peso molecolare: 195,2 g/mol Punto di fusione: 285…287 °C |
MDL Number:
MFCD00804727 CAS Number: 6325-91-3 |
Risultati del test di specificazione
Appearance | Light yellow to Brown powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...