Stampa…

2,4-Dihydroxy-3-nitro-6-picoline ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2,4-Dihydroxy-6-methyl-3-nitropyridine, 4-Hydroxy-6-methyl-3-nitropyridin-2(1H)-one, 4-Hydroxy-6-methyl-3-nitro-2-pyridone

H0921-25G H0921-5G
TCIAH0921-25GEA 103 EUR
TCIAH0921-25G TCIAH0921-5G
2,4-Dihydroxy-3-nitro-6-picoline ≥98.0% (by GC, titration analysis)
2,4-Dihydroxy-3-nitro-6-picoline
Formula: C₆H₆N₂O₄
Peso molecolare: 170,12 g/mol
MDL Number: MFCD00167612
CAS Number: 4966-90-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b7a754f9c2eb6495e43006818e63d061de5ee9c3&vid=b7a754f9c2eb6495e43006818e63d061de5ee9c3&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Solubility in N,N-DMF very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR