2-Hydroxy-3-nitro-4-picoline ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
4-Methyl-3-nitro-2-pyridone, 2-Hydroxy-4-methyl-3-nitropyridine
H1170-1G
H1170-5G
TCIAH1170-1GEA
24
EUR
TCIAH1170-1G
TCIAH1170-5G
2-Hydroxy-3-nitro-4-picoline ≥98.0% (by GC, titration analysis)
2-Hydroxy-3-nitro-4-picoline
Formula:
C₆H₆N₂O₃ Peso molecolare: 154,13 g/mol Punto di fusione: 234°C |
MDL Number:
MFCD00010689 CAS Number: 21901-18-8 |
Risultati del test di specificazione
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...