Stampa…

4-Iodoimidazole ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 4(5)-Iodoimidazole, 5-Iodo-1H-imidazole, 4-Iodo-1H-imidazole

I0812-1G I0812-5G
TCIAI0812-1GEA 41 EUR
TCIAI0812-1G TCIAI0812-5G
4-Iodoimidazole ≥98.0% (by HPLC, titration analysis)
4-Iodoimidazole
Formula: C₃H₃IN₂
Peso molecolare: 193,97 g/mol
Punto di fusione: 137…138 °C
MDL Number: MFCD01632213
CAS Number: 71759-89-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bef58231c752a03f9487af79e155893a1d82f282&vid=bef58231c752a03f9487af79e155893a1d82f282&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 136.0 to 140.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR