3,5-Bis(trifluoromethyl)benzenesulphonyl chloride ≥98.0% (by GC, titration analysis)
Fornitore: TCI
|
Pericolo
|
Formula:
C₈H₃ClF₆O₂S Punto di fusione: 36…38 °C |
MDL Number:
MFCD00014725 CAS Number: 39234-86-1 EINECS: 254-371-0 UN: 3261 ADR: 8,II |
|
Risultati del test di specificazione
Appearance | White to Orange to Green powder to lump |
Purity (GC) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...