Methyl heptafluorobutyrate ≥97.0% (by GC)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
Methyl 2,2,3,3,4,4,4-heptafluorobutanoate, Methyl perfluorobutyrate
H1033-25G
H1033-5G
TCIAH1033-25GEA
112
EUR
TCIAH1033-25G
TCIAH1033-5G
Methyl heptafluorobutyrate ≥97.0% (by GC)
Methyl heptafluorobutyrate
Formula:
C₅H₃F₇O₂ Peso molecolare: 228,07 g/mol Punto di ebollizione: 81°C Punto di fusione: –86 °C Densità: 1,472 g/cm³ (20 °C) |
MDL Number:
MFCD00000433 CAS Number: 356-24-1 EINECS: 206-600-0 UN: 3272 ADR: 3,II |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...