Stampa…

4-Fluoro-m-anisaldehyde ≥96.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 4-Fluoro-3-methoxybenzaldehyde, 4-Fluoro-3-anisaldehyde

F0401-25G F0401-5G F0401-1G
TCIAF0401-25GEA 205 EUR
TCIAF0401-25G TCIAF0401-5G TCIAF0401-1G
4-Fluoro-m-anisaldehyde ≥96.0% (by GC)
4-Fluoro-m-anisaldehyde
Formula: C₈H₇FO₂
Peso molecolare: 154,14 g/mol
MDL Number: MFCD00143320
CAS Number: 128495-46-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=06273e0bef5cb3f3c31eb6c174419094b615210e&vid=06273e0bef5cb3f3c31eb6c174419094b615210e&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 96.0 %
Melting point 58.0 to 62.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR