Stampa…

Bis(4-nitrophenyl)carbonate ≥98.0% (by HPLC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Bis(4-nitrophenyl) carbonate

C1481-25G C1481-5G
TCIAC1481-25GEA 100 EUR
TCIAC1481-25G TCIAC1481-5G
Bis(4-nitrophenyl)carbonate ≥98.0% (by HPLC)
Bis(4-nitrophenyl)carbonate
Formula: C₁₃H₈N₂O₇
Peso molecolare: 304,22 g/mol
MDL Number: MFCD00007322
CAS Number: 5070-13-3
EINECS: 225-775-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=77a7ae9563a0de0dd6b3d11f3977c8b46745e4e3&vid=77a7ae9563a0de0dd6b3d11f3977c8b46745e4e3&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 140.0 to 144.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR