Stampa…

4-Nitrophenyl acetate ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: P-Acetoxynitrobenzene, 1-Acetoxy-4-nitrobenzene

A0040-25G A0040-100G
TCIAA0040-25GEA 126 EUR
TCIAA0040-25G TCIAA0040-100G
4-Nitrophenyl acetate ≥98.0% (by GC)
4-Nitrophenyl acetate
Formula: C₈H₇NO₄
Peso molecolare: 181,15 g/mol
Punto di fusione: 76…79 °C
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00007326
CAS Number: 830-03-5
EINECS: 212-593-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a605faceb75b90112b338187ee4ac74bf4963365&vid=a605faceb75b90112b338187ee4ac74bf4963365&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 77.0 to 80.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR