Mucic acid ≥97.0% (by titrimetric analysis)
Fornitore: TCI
|
Attenzione
|
Formula:
C₆H₁₀O₈ Punto di fusione: 220…225 °C Densità: 1,79 g/cm³ (20 °C) |
MDL Number:
MFCD00004239 CAS Number: 526-99-8 EINECS: 208-404-0 Merck Index: 14,04331 |
|
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | min. 97.0 % |
Solubility in Sodium hydroxide solution | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...