Stampa…

Trimesic acid trimethyl ester ≥98.0% (by GC)

Fornitore: TCI

T2707-25G T2707-5G
TCIAT2707-25GEA 99 EUR
TCIAT2707-25G TCIAT2707-5G
Trimesic acid trimethyl ester ≥98.0% (by GC)
Trimesic acid trimethyl ester
Formula: C₁₂H₁₂O₆
Punto di ebollizione: 230°C
Punto di fusione: 146,5…147 °C
MDL Number: MFCD00008434
CAS Number: 2672-58-4
EINECS: 220-215-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=89fc7b50dd7d16085743fe120ee8c955c2f7cca6&vid=89fc7b50dd7d16085743fe120ee8c955c2f7cca6&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 144.0 to 148.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR