Methyl methoxyacetate ≥97.0% (by GC)
Fornitore: TCI
|
Attenzione
|
M0863-25ML
M0863-500ML
TCIAM0863-25MLEA
17
EUR
TCIAM0863-25ML
TCIAM0863-500ML
Methyl methoxyacetate ≥97.0% (by GC)
Methyl methoxyacetate
Formula:
C₄H₈O₃ Punto di ebollizione: 132°C Densità: 1,0579 g/cm³ (15 °C) Flash Pt: 35°C |
MDL Number:
MFCD00008451 CAS Number: 6290-49-9 EINECS: 228-539-9 UN: 3272 ADR: 3,III |
|
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...