Ethyl-3-phenylpropionate ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
Ethyl 3-Phenylpropionate, Ethyl hydrocinnamate, 3-Phenylpropionic acid ethyl ester, Ethyl dihydrocinnamate, Hydrocinnamic acid ethyl ester
P1304-25G
P1304-250G
TCIAP1304-25GEA
25
EUR
TCIAP1304-25G
TCIAP1304-250G
Ethyl-3-phenylpropionate ≥98.0% (by GC)
Ethyl-3-phenylpropionate
Formula:
C₁₁H₁₄O₂ Peso molecolare: 178,23 g/mol Punto di ebollizione: 248°C Densità: 1,0343 g/cm³ (0 °C) Flash Pt: 108°C |
MDL Number:
MFCD00009206 CAS Number: 2021-28-5 EINECS: 217-966-6 |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...