(4-Pyridyl)acetic acid ethyl ester ≥98.0% (by GC, titration analysis)
Fornitore: TCI
|
Attenzione
|
P1399-25G
P1399-5G
TCIAP1399-25GEA
259
EUR
TCIAP1399-25G
TCIAP1399-5G
(4-Pyridyl)acetic acid ethyl ester ≥98.0% (by GC, titration analysis)
(4-Pyridyl)acetic acid ethyl ester
Formula:
C₉H₁₁NO₂ |
MDL Number:
MFCD00051830 CAS Number: 54401-85-3 EINECS: 259-150-2 |
|
Risultati del test di specificazione
Appearance | White to Almost white clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...