Stampa…

Naphthol AS ≥97.0% (by HPLC, titration analysis)

Fornitore: TCI
Attenzione

H0313-25G H0313-500G
TCIAH0313-25GEA 16 EUR
TCIAH0313-25G TCIAH0313-500G
Naphthol AS ≥97.0% (by HPLC, titration analysis)
Naphthol AS
Formula: C₁₇H₁₃NO₂
Punto di ebollizione: 494 °C (1013 hPa)
Punto di fusione: 252 °C
MDL Number: MFCD00004096
CAS Number: 92-77-3
EINECS: 202-188-1
UN: 3077
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=413aec357d93b30168783dd64063c59c1fbcfaca&vid=413aec357d93b30168783dd64063c59c1fbcfaca&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) min. 97.0 %
Melting point 247.0 to 251.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR