Baicalein ≥98.0% (by titrimetric analysis)
Fornitore: TCI
|
Attenzione
|
Formula:
C₁₅H₁₀O₅ Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00017459 CAS Number: 491-67-8 Merck Index: 14,00942 |
|
Risultati del test di specificazione
Appearance | Light yellow to Amber to Dark green powder to crystal |
Purity (HPLC) | min. 95.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...