6-Hydroxyflavone ≥98.0% (by HPLC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
6-Hydroxy-2-phenylchromen-4-one, 6-Hydroxy-2-phenyl-4H-chromen-4-one, 6-Hydroxy-2-phenyl-4H-benzo[b]pyran-4-one, 6-Hydroxy-2-phenyl-4-benzopyrone, 6-Hydroxy-2-phenyl-4H-1-benzopyran-4-one
H0851-1G
H0851-5G
TCIAH0851-1GEA
34
EUR
TCIAH0851-1G
TCIAH0851-5G
6-Hydroxyflavone ≥98.0% (by HPLC)
6-Hydroxyflavone
Formula:
C₁₅H₁₀O₃ Peso molecolare: 238,24 g/mol Punto di ebollizione: 395 °C (1013 hPa) Punto di fusione: 239 °C |
MDL Number:
MFCD00017329 CAS Number: 6665-83-4 EINECS: 229-704-8 |
Risultati del test di specificazione
Appearance | Light yellow to Yellow to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 97.0 % |
Solubility in hot Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...