Stampa…

4-Acetoxybenzoic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

A0885-25G A0885-500G
TCIAA0885-25GEA 29 EUR
TCIAA0885-25G TCIAA0885-500G
4-Acetoxybenzoic acid ≥98.0% (by GC, titration analysis)
4-Acetoxybenzoic acid
Formula: C₉H₈O₄
Peso molecolare: 180,16 g/mol
MDL Number: MFCD00002540
CAS Number: 2345-34-8
EINECS: 219-066-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bdbde010130fbb872234841351dfd4c613562281&vid=bdbde010130fbb872234841351dfd4c613562281&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 190.0 to 196.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR