α-Naphthoflavone ≥98.0% (by HPLC)
Fornitore: TCI
Formula:
C₁₉H₁₂O₂ Punto di ebollizione: 431 °C (1013 hPa) Punto di fusione: 156…157 °C |
MDL Number:
MFCD00004985 CAS Number: 604-59-1 EINECS: 210-071-1 |
|
Risultati del test di specificazione
Appearance | White to Yellow to Orange powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Melting point | 154.0 to 158.0 deg-C |
Solubility in hot EtOH | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...