4,5-Dicyanoimidazole ≥98.0% (by HPLC, total nitrogen)
Fornitore: TCI
|
Pericolo
|
Synonyms:
4,5-Dicyano-1H-imidazole, Imidazole-4,5-dicarbonitrile
Formula:
C₅H₂N₄ Punto di fusione: 176 °C Densità: 1,458 g/cm³ (20 °C) |
MDL Number:
MFCD00005194 CAS Number: 1122-28-7 EINECS: 214-344-6 UN: 3439 ADR: 6.1,III |
|
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Solubility in hot Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...