Stampa…

2-Chloro-4-methyl-3-nitropyridine ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2-Chloro-3-nitro-4-picoline

C2012-1G C2012-5G
TCIAC2012-1GEA 20 EUR
TCIAC2012-1G TCIAC2012-5G
2-Chloro-4-methyl-3-nitropyridine ≥98.0% (by GC)
2-Chloro-4-methyl-3-nitropyridine
Formula: C₆H₅ClN₂O₂
Peso molecolare: 172,57 g/mol
MDL Number: MFCD00012347
CAS Number: 23056-39-5
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e049d0faad8d284e1f448539f639a558de22ec1a&vid=e049d0faad8d284e1f448539f639a558de22ec1a&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 51.0 to 55.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR