2-Chloro-4-(trifluoromethyl)benzenesulphonyl chloride ≥98.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
2-Chloro-4-(trifluoromethyl)benzene-1-sulphonyl chloride
C1790-1G
C1790-5G
TCIAC1790-1GEA
56
EUR
TCIAC1790-1G
TCIAC1790-5G
2-Chloro-4-(trifluoromethyl)benzenesulphonyl chloride ≥98.0% (by titrimetric analysis)
2-Chloro-4-(trifluoromethyl)benzenesulphonyl chloride
Formula:
C₇H₃Cl₂F₃O₂S Peso molecolare: 279,07 g/mol Punto di ebollizione: 70 - 71 °C (0,5 -rr) |
MDL Number:
MFCD00052912 CAS Number: 175205-54-6 UN: 3265 ADR: 8,III |
Risultati del test di specificazione
Appearance | Light orange to Yellow to Green clear liquid |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...