Stampa…

4-Chloro-3-(trifluoromethyl)phenol ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2-Chloro-5-hydroxybenzotrifluoride

C1516-1G C1516-5G
TCIAC1516-1GEA 27 EUR
TCIAC1516-1G TCIAC1516-5G
4-Chloro-3-(trifluoromethyl)phenol ≥98.0% (by GC)
4-Chloro-3-(trifluoromethyl)phenol
Formula: C₇H₄ClF₃O
Peso molecolare: 196,56 g/mol
Punto di ebollizione: 122°C
MDL Number: MFCD00042525
CAS Number: 6294-93-5
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bab85afd0ab0600854093d7827f1f22524c05010&vid=bab85afd0ab0600854093d7827f1f22524c05010&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 42.0 to 46.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR