2-Amino-5-chlorobenzophenone ≥98.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
1-Amino-4-chloro-2-(phenylmethanoyl)benzene, (2-Amino-5-chlorophenyl)(phenyl)methanone
A0079-25G
A0079-500G
TCIAA0079-25GEA
31
EUR
TCIAA0079-25G
TCIAA0079-500G
2-Amino-5-chlorobenzophenone ≥98.0% (by titrimetric analysis)
2-Amino-5-chlorobenzophenone
Formula:
C₁₃H₁₀ClNO Peso molecolare: 231,68 g/mol Punto di ebollizione: 424 °C (1013 hPa) Punto di fusione: 96…99 °C |
MDL Number:
MFCD00007839 CAS Number: 719-59-5 EINECS: 211-949-7 |
Risultati del test di specificazione
Appearance | Light yellow to Yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 96.0 to 100.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...