1-Ethylpyridinium chloride ≥98.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
1-Ethylpyridin-1-ium chloride
E0544-25G
E0544-5G
TCIAE0544-25GEA
288
EUR
TCIAE0544-25G
TCIAE0544-5G
1-Ethylpyridinium chloride ≥98.0% (by titrimetric analysis)
1-Ethylpyridinium chloride
Formula:
C₇H₁₀ClN Peso molecolare: 143,62 g/mol |
MDL Number:
MFCD03093293 CAS Number: 2294-38-4 |
Risultati del test di specificazione
Appearance | White to Blue powder to lump |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...