Stampa…

4-Bromo-3-methylbenzenesulphonyl chloride ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: 4-Bromo-3-methylbenzene-1-sulphonyl chloride

B2343-1G B2343-5G
TCIAB2343-1GEA 56 EUR
TCIAB2343-1G TCIAB2343-5G
4-Bromo-3-methylbenzenesulphonyl chloride ≥98.0% (by GC, titration analysis)
4-Bromo-3-methylbenzenesulphonyl chloride
Formula: C₇H₆BrClO₂S
Peso molecolare: 269,55 g/mol
Punto di fusione: 61 °C
MDL Number: MFCD03094696
CAS Number: 72256-93-0
UN: 3261
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d0e59054a7c740a6e3de2ce59806407bb76e89ce&vid=d0e59054a7c740a6e3de2ce59806407bb76e89ce&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Melting point 57.0 to 62.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR