Stampa…

2-Bromo-4-methoxyphenol ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 3-Brom-4-hydroxyanisole

B3135-25G B3135-5G
TCIAB3135-25GEA 179 EUR
TCIAB3135-25G TCIAB3135-5G
2-Bromo-4-methoxyphenol ≥98.0% (by titrimetric analysis)
2-Bromo-4-methoxyphenol
Formula: C₇H₇BrO₂
Punto di ebollizione: 115°C
MDL Number: MFCD02113712
CAS Number: 17332-11-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0006b972d3c05c34ba70f597fc12c5937b52a2f&vid=b0006b972d3c05c34ba70f597fc12c5937b52a2f&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 96.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 39.0 to 45.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR