Stampa…
Methyl-2-bromo-4-nitrobenzoate ≥98.0% (by GC)
Fornitore: TCI
Attenzione
Synonyms: Methyl 2-bromo-4-nitrobenzoate, 2-Bromobenzoic acid methyl ester
M2123-1G M2123-5G
TCIAM2123-1GEA 17 EUR
TCIAM2123-1G TCIAM2123-5G
Methyl-2-bromo-4-nitrobenzoate ≥98.0% (by GC)
Methyl-2-bromo-4-nitrobenzoate
Formula: C₈H₆BrNO₄
Peso molecolare: 260.04 g/mol
MDL Number: MFCD07779271
CAS Number: 100959-22-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f9ff710455a53303e3d0ca5efe0c4b56587c4b26&vid=f9ff710455a53303e3d0ca5efe0c4b56587c4b26&prid=web&clid=SAPDAM
Visualizza i codici
Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 82.0 to 86.0 deg-C