5-Bromopentyl acetate ≥95.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
5-Bromoamyl acetate, Acetic acid 5-bromoamyl ester, Acetic acid 5-bromopentyl ester
A1195-100G
A1195-25G
A1195-5G
TCIAA1195-100GEA
161
EUR
TCIAA1195-100G
TCIAA1195-25G
TCIAA1195-5G
5-Bromopentyl acetate ≥95.0% (by GC)
5-Bromopentyl acetate
Formula:
C₇H₁₃BrO₂ Peso molecolare: 209,08 g/mol Punto di ebollizione: 113°C |
MDL Number:
MFCD00039204 CAS Number: 15848-22-3 EINECS: 239-961-8 |
Risultati del test di specificazione
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...