Stampa…

(±)-2-Hexanol ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: (±)-Methyl butyl carbinol, (±)-Butyl methyl carbinol, (±)-2-Hydroxy hexane

H0131-25ML H0131-500ML H0131-100ML
TCIAH0131-25MLEA 43 EUR
TCIAH0131-25ML TCIAH0131-500ML TCIAH0131-100ML
(±)-2-Hexanol ≥98.0% (by GC)
(±)-2-Hexanol
Formula: C₆H₁₄O
Peso molecolare: 102,18 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00004585
CAS Number: 626-93-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5a6e95f48f5a8567d32c0e14217905109f543637&vid=30d09f2f591bc9aa534924eb9b26d59e117b13b6&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR