Stampa…

Sodium cyclamate ≥98.0% (by total nitrogen basis)

Fornitore: TCI
Attenzione

C0487-25G C0487-500G
TCIAC0487-25GEA 16 EUR
TCIAC0487-25G TCIAC0487-500G
Sodium cyclamate ≥98.0% (by total nitrogen basis)
Sodium cyclamate
Formula: C₆H₁₂NNaO₃S
Punto di fusione: 265 °C
MDL Number: MFCD00003827
CAS Number: 139-05-9
EINECS: 205-348-9
Merck Index: 14,02703
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b1d84ec481c98cca7c691feb76c32931e4f02907&vid=b1d84ec481c98cca7c691feb76c32931e4f02907&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR